Detergents, proteomic grade
Lieferant: G-Biosciences
DG521
DG521EA 397 EUR
DG521
Detergents, proteomic grade
Reinigungsmittel Waschmittel und Oberflächenreiniger
This variety pack contains non ionic proteomic grade detergent solutions, zwitterionic and non-detergent sulphobetaines for trial and optimisation.
Lieferumfang: The pack contains one 10 ml vial of 10% aqueous solutions of Triton® X-100, Triton® X-114, Tween® 20, Tween® 80, Nonidet® P-40 Substitute, Brij® 35, Brij® 58 and 1 g of CHAPS, NDSB 201.