Intelsius® Biomailer
Lieferant: DGP INTELSIUS
BM003
DGINBM003EA 1080 EUR
DGINBM003
Intelsius® Biomailer
Versandtaschen und Transportbehälter
The Intelsius® Biomailer is designed for use within national and global postal networks shipping small specimens, such as blood vials and slides.
  • Rigid plastic case
  • Ship small specimens
  • National and global postal mailer
  • Supplied with plastic pouch and absorbent
  • Multi-use device
The rigid plastic case ensures that samples are protected during transit, and allows the system to be refurbished and used again, offering both a cost saving and environmental benefit.
The weight is 0,4 lbs.
Standards: ISTA Certified Class 6,2, Category B.
Bestellinformation: Includes A5 pathopouch (95 kPa secondary), 4 bay absorbent, protective sleeve, UN3373 biological substance category B label, and security seal label.