Kernechtrot

Lieferant: DIANOVA

Synonyme: Natrium-1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracen-3-sulfonat, Heliechtrubin

15904
DIAN15904EA 0 EUR
DIAN15904
Kernechtrot
Kernechtrot
Formel: C₁₄H₈NNaO₇S
Molecular Weight: 357.28 g/mol
Schmelzpunkt: 300
Lagertemperatur: Raumtemperatur
MDL: MFCD00078493
CAS-Nummer: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR