Cholesterin FS 10
Lieferant: DIAGNOSTIC SYSTEMS DIASYS
Synonyme:
5-Cholesten-3β-ol, 3β-Hydroxy-5-cholesten, Cholesterol, Cholest-5-en-3β-ol
113009910917
DIAS113009910917EA
0
EUR
DIAS113009910917
Cholesterin FS 10
Cholesterin
Formel:
C₂₇H₄₆O Molecular Weight: 386,66 g/mol Dichte: 1,0050 g/ml at 20 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00003646 CAS-Nummer: 57-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...