Temperature Loggers
Lieferant: DICKSON
SM320
DICKSM320EA 717 EUR
DICKSM320
Temperature Loggers
Datenlogger
These temperature loggers are designed for monitoring, recording and evaluating temperature of critical equipment, computers, HVAC/R systems, incubators, freezers, autoclaves, baths and cleanrooms. They have a high/low audible alarms.
  • Large four-digit LCD features
  • Flash memory option for easy data transfer
  • USB enabled fast downloading
  • Min/max display
  • Temperature model and remote probe model
  • Super high speed USB 2.0 connectivity
  • Download data faster when compared to using standard serial port
A memory card reader kit and removeable Flash memory storage cards are available for different units. The reader kit includes a USB memory card reader and a 128 MB, 1 Gb flash memory card. They require the use of DicksonWare™ software which must be purchased separately.
Lieferumfang: Supplied with four AA alkaline batteries.