Optical equipment to demonstrate the law of polarisation
Lieferant: DIDALAB
POD069125 POD060910 POD060920 POD060930 POD060955 POD060965
DIDAPOD069125EA 0 EUR
DIDAPOD069125 DIDAPOD060910 DIDAPOD060920 DIDAPOD060930 DIDAPOD060955 DIDAPOD060965
Optical equipment to demonstrate the law of polarisation
Lehrmittel Physik-Lehrmittel Lehrmittel für die Optik
This comprehensive optical pack allows you to implement and study the basic principles of polarisation of light.
  • Study the state of polarisation of sources
  • Analyse the effects of various polarisation retardation plates
Bestellen
TECHNISCHE DATEN
Environmentally Preferable