CCD camera, POF 010 300
Lieferant: DIDALAB
POF010300
DIDAPOF010300EA 306 EUR
DIDAPOF010300
CCD camera, POF 010 300
Kameras
This Caliens camera enables measuring very precise distances.
  • CCD sensor, 2048 pixels
  • User-friendly software, Windows compatible
  • Power supplied via USB port (connected to a PC)
Lieferumfang: Supplied with Ø 10 mm rod for use with optical benches, software and USB cable.