Cable for CCD sensor
Lieferant: DIDALAB
POF010610
DIDAPOF010610EA 0 EUR
DIDAPOF010610
Cable for CCD sensor
Kameras
This cable retrieves the “raw” signals of the sens interface. It helps to explain the principle of CCD photosensitive sensors and show the relation between the light received by the sensor and the electrical measure.