Sulphate reducing bacteria test kit
Lieferant: DIMANCO
SRB
DIMASRBEA
30.9
EUR
DIMASRB
Sulphate reducing bacteria test kit
Mikrobiologische Testsysteme
Tests zur Überprüfung von Mirkoorganismen
Dipslide test for sulphate-reducing bacteria.
The dip and contact slides have many special features. The unique design gives the slide considerable flexibility for maximum accuracy.