Sulphate reducing bacteria test kit
Lieferant: DIMANCO
SRB
DIMASRBEA 30.9 EUR
DIMASRB
Sulphate reducing bacteria test kit
Mikrobiologische Testsysteme Tests zur Überprüfung von Mirkoorganismen
Dipslide test for sulphate-reducing bacteria.
The dip and contact slides have many special features. The unique design gives the slide considerable flexibility for maximum accuracy.