Optical DO Sensor Cable
Lieferant: DISTEK
2700-0418
DISI2700-0418EA 0 EUR
DISI2700-0418
Optical DO Sensor Cable
Pumpen Peristaltikpumpen
Optical DO sensor cable that will work only with the optical DO sensors such as the Hamilton VisiFerm ECS optical DO sensor.