Colour-coded Tough-Tags® labels
Lieferant: DIVERSIFIED BIOTECH
COLB-1000 COLR-1000 COLG-1000 COLY-1000 COLW-1000
DIVECOLB-1000EA 144 EUR
DIVECOLB-1000 DIVECOLR-1000 DIVECOLG-1000 DIVECOLY-1000 DIVECOLW-1000
Colour-coded Tough-Tags® labels
Etiketten
These Tough-Tags® polyester colour-coded labels make it easy to differentiate samples and identify what you need.
  • Temperature resistant from −80 to +90 °C
  • Translucent writing area keeps contents visible
  • Set consists of a colour-coded rectangular label and spot
The labels are repositionable for up to thirty minutes as the adhesive strengthens over time.