Anti-microbial benchtop mats, Silver Bioguard
Lieferant: DIVERSIFIED BIOTECH
SLVR-3300
DIVESLVR-3300EA 0 EUR
DIVESLVR-3300
Anti-microbial benchtop mats, Silver Bioguard
Oberflächenschutz Oberflächenschutz
These anti-microbial mats contain silver nanoparticles to create contamination free work surface.
  • Nanoscale Silver particles bind with proteins to neutralise 99,9% of common microbes
  • Ensure consistent sample reproduction and results by significantly minimising workspace contamination
  • Silver is used extensively in medical applications where microbial contamination is a concern
  • Backed with anti-skid coating, perfect for the benchtop or culture hood