Thermal transfer Tough-Tags® labels
Lieferant: DIVERSIFIED BIOTECH
THER-1137 THER-1094
DIVETHER-1137EA 362 EUR
DIVETHER-1137 DIVETHER-1094
Thermal transfer Tough-Tags® labels
Etiketten
These Tough-Tags® labels are used to print crisp, clear, professional quality labels with thermal transfer printers.
  • Temperature resistant from -40 to +150 °C
  • Solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
  • Resists degradation from scuffing, chemicals, moisture and wide temperature fluctuations
The labels are coated for use with a resin ribbon that provides the best performance under extreme conditions.