These Wash-Off labels and adhesive completely dissolve in any water temperature in under 30 seconds. They readily accept laser printing for quick and easy labeling.
FDA-compliant for indirect food contact
Autoclavable, resist exposure to steam at 121 °C
Storage at room temperature to moderate freezer temperatures
Fits a variety of bottles, flasks, beakers and other laboratory containers.