Identification cards, 'Well Orienter'
Lieferant: DIVERSIFIED BIOTECH
WELL-1000
DIVEWELL-1000EA 114 EUR
DIVEWELL-1000
Identification cards, 'Well Orienter'
Titerplatten-Ständer und Well-Orientierungshilfen
This Well Orienter allows easy pipetting and identification of samples in microplates. The thin plastic card that slips under the well plate provides sharp, alphanumeric characters to identify each sample in 96-well microplates.
  • Alphanumeric coding