Cleaning disinfectant, oxivir plus spray
Lieferant: Diversey
100829236
DIVY100829236EA 107 EUR
DIVY100829236
Cleaning disinfectant, oxivir plus spray
Reinigungsmittel Desinfektionsmittel
Oxivir plus spray is a concentrated washing and disinfectant prepared on active oxygen base AHP. Ideal for textiles and hard surfaces.
  • Hospital grade disinfectant
  • Based on proprietary hydrogen peroxide technology (AHP®)
  • Disinfects in 5 minutes at 1:40 dilution
  • Excellent surface safety
  • Kills MRSA ,VRE and Norovirus and Canine Parvovirus