SURE® Ice and shake is a concentrated detergent disinfectant suitable for the cleaning, disinfection and milkstone removal of shake, ice machines and other beverage equipment found in food service premises.
- Saves time; cleaning, disinfection and milkstone removal is done with one single product in one application
- Very effective against a wide range of microorganisms; bactericidal and yeasticidal activities
- Contains plant based actives from renewable sources decomposed by natural processes
- No environmental hazard classifications, no safety warnings for product in use for user
- No fragrance nor dye, natural odour present due to plant based ingredients
SURE® Ice & Shake is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance. Bottles/cans suitable for recycling.
Standards: OECD 301B. Cradle to cradle certification at gold level.