Oxivir Excel® CE Cleaner and Disinfectant
Lieferant: Diversey
100941438
DIVY100941438EA 156 EUR
DIVY100941438
Oxivir Excel® CE Cleaner and Disinfectant
Reinigungsmittel Desinfektionsmittel
Highly concentrated one step cleaner disinfectant with broad spectrum efficacy and short contact times. Based on Diversey Care patented Accelerated Hydrogen Peroxide formulation Oxivir Excel can be used on non invasive medical devices.
  • Full virucidal claim in just 30 seconds
  • Fast kill claims for a broad spectrum of viruses, bacteria and yeasts
  • Effective Cleaner
  • Fragrance free
  • No rinsing required 
  • Safe for the environment, the users and the patients, since the active ingredient breaks down into water and oxygen
  • Does not contain chlorine or alcohol
Benefits:
Broad efficacy provides the highest levels of disinfection. Extensively tested against European norms and effective against multiple pathogens of concern, carries a full virucidal claim including Poliovirus, Adenovirus, and Murine norovirus, considered effective against all enveloped viruses such as: HIV, HBV, HCV, Influenza, Vaccinia virus and Coronavirus
Short contact times keep the surface wet thus ensuring disinfectant efficacy 
Suitable for use in high risk areas in healthcare
Immersion method:
  • Dilute the concentrated liquid as required (see microbiological data table below).
  • Remove contamination with a damp cloth (gross soils).
  • Immerse the medical device fully for the required contact time (see microbiological data table below).
  • After the contact time, remove the medical device out, wipe dry or allow to air dry.
Wiping/mopping method:
  • Dilute the concentrated liquid as required (see microbiological data table below).
  • Apply the solution to the medical device surface by spraying.
  • Wipe the medical device surface using a clean cloth and ensure it remains wet during the contact time indicated in the table below.
  • After the contact time, wipe the surface dry or allow to air dry.
Weight: 10,65 kg (Net weight), 11,33 kg (Gross weight)
Dimension (W×L×H): 198×287×309 mm (Traded unit), 135×185×283 mm (Consumer unit)