Sporicide wipes, Oxivir®
Lieferant: Diversey
100941885
DIVY100941885EA 177 EUR
DIVY100941885
Sporicide wipes, Oxivir®
Wischtücher Wisch- und Handtücher, Einweg Desinfektionstücher
Oxivir® Sporicide wipes are a one-step disinfectant cleaner effective against Clostridioides difficile and Bacillus subtilis.
  • Sporicidal, Bactericidal, Virucidal, Yeasticidal, Fungicidal, in as little as 1 minute
  • Short contact times keep the surface wet, thus ensuring disinfectant efficacy
  • No rinsing required
  • Active breaks down into water and oxygen - no active residues left on the surface
  • Does not contain chlorine or alcohol
  • Does not cause respiratory irritation
  • Suitable for use in high risk areas in Healthcare, including ICU, NICU, patient and isolation rooms
Based on Accelerated Hydrogen peroxide formulation Oxivir® Sporicide Wipes can be used on hard, non-porous surfaces and non-invasive medical devices.
Standards: Efficacy proven by compliance with: EN 13727, EN 13624, EN 13697, EN 14476, EN 13704, EN 14348 and EN 16615. The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.
Bitte beachten Gloves should be worn when using Oxivir Sporicide wipe. Do not mix with other products. Do not use on water- or acid-sensitive surfaces.
*Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904