TASKI® Jontec No1 F1c Heavy-Duty Floor Stripper - No Rinse
Lieferant: Diversey
101102722
DIVY101102722EA 39.5 EUR
DIVY101102722
TASKI® Jontec No1 F1c Heavy-Duty Floor Stripper - No Rinse
Reinigungsmittel Waschmittel und Oberflächenreiniger
Heavy duty stripper for effective removal of floor emulsions and maintenance films from alkali resistant hard floors.
  • Highly alkaline
  • High wetting and emulsifying power
  • Fast and easy emulsion removal without rinsing
Unlike conventional strippers, its revolutionary formulation enables floors to be ready for re-coating without the need to rinse, reducing normal stripping time by up to 50%.