Profile solution Detergens, zum Reinigen, chloriert
Lieferant: Diversey
Profile is a medium hard water tolerant broad spectrum, heavy duty, low foam chlorinated alkaline detergent used in CIP and spray washing applications. Profile has excellent protein removal properties. Profile is used in the meat processing industry for mechanical soak cleaning and tray washing applications.
- Profile provides excellent detergency for protein removal in spray washing and CIP applications in the food and beverage industries
- Profile is medium hard water tolerant
- Profile is suitable for CIP under conditions of high turbulence
- Medium hard water tolerant, ensuring scale free equipment and improved operational efficiency
- Low foaming under high turbulence making it easier to rinse and improving cleaning efficiency
Profile is used at 0.5 to 1.5% w/w (0.4 to 1.3% v/v) at ambient to 50°C for soak applications and at 50 °C for CIP and spray washing applications. Profile is used at 40 to 50 °C for 10 to 20 minutes for CIP of milk tanks, tray washing and packing machines. Profile should be rinsed thoroughly from all surfaces after cleaning, to ensure potential staining is eliminated.