Alkaline detergent, powder, Suma Strip K9
Lieferant: Diversey
7510952
DIVY7510952EA 280 EUR
DIVY7510952
Alkaline detergent, powder, Suma Strip K9
Reinigungsmittel Waschmittel und Oberflächenreiniger
Highly alkaline powdered detergent for periodical removal of starch residues from dishes or heavily soiled surfaces in food areas.
  • Low foaming properties to improve cleaning efficiency
  • Inhibits scale build up