TASKI Sani Antikalk Heavy Duty Washroom Cleaners, W3e
Lieferant: Diversey
7513263
DIVY7513263EA 22.5 EUR
DIVY7513263
TASKI Sani Antikalk Heavy Duty Washroom Cleaners, W3e
Reinigungsmittel Waschmittel und Oberflächenreiniger
Acid-based sanitary cleaner and limescale remover for acid resistant hard surfaces.
  • Sulphamic acid based
  • Contains a corrosions inhibitor
  • Strong descaling properties
  • Unique, patented Odour Neutralizing Technology (O.N.T.)
Effective removal of stubborn calcium/limescale and of common sanitary soil, e.g. body fat and soap residues. Perfectly fit for efficient full surface descaling of floors, walls and other washroomsurfaces. Ready to use, streak free cleaning. Leaves a pleasant, fresh fragrance.
Spray method:
- Spray the solution on a damp cloth and wipe
- Use a sponge pad to remove stubborn soil
- Rinse or replace the cloth regularly