Courier Case
Lieferant: DELTA T
B100040
DLTTB100040EA 887 EUR
DLTTB100040
Courier Case
Versandtaschen und Transportbehälter
Courier case suitable for 3,5 pairs of TempShell elements or combinations of elements and TempShell frames. Robust plastic case with vacuum insulation, lock with 'butterfly' closures. Shoulder strap, carrying handle on the lid, sealable and washable plastic interior.
Weight: 5,98 kg.