High Precision Reference Thermometer, P795
Lieferant: TFA DOSTMANN
5000-0795
DOST5000-0795EA 1740 EUR
DOST5000-0795
High Precision Reference Thermometer, P795
Thermometer Digitalthermometer
An instrument which meets the demands. The 2-channel-resistance thermometer automatically detects our Pt100-smartprobes with integrated EEprom. Thereby the calibration characteristic of each probe is transferred automatically to the instrument.
  • 2-channel-instruments, for PT100 according EN 60751
  • Simultaneous display of two measured values or differential measurement
  • Recording maximum, minimum, hold and average values (average over user selectable time)
  • Integrated calibration function (acc. EN60751 up to 14 points)
  • High measuring accuracy (+0,010 °C)
  • Includes USB-cable
  • Data Logger function for up to 6000 measurements
  • Optional DAKKS or ISO-system calibration certification
The thermometer will achieve a system accuracy of ±0,015 K (instrument + probe). Both measuring channels can be easily seen on the large LC display. The instrument contains numerous functions for the measurement visualisation. Various integrated calibration functions, including the intelligent EEprom-probes, takes this compact instrument to a high performance class, making it ideally suitable as a primary reference standard.