Infrared thermometer, Slim Flash
Lieferant: TFA DOSTMANN
5020-0325
DOST5020-0325EA 46.5 EUR
DOST5020-0325
Infrared thermometer, Slim Flash
Thermometer Digitalthermometer
These infrared thermometers are for contact-free measuring of surface temperature in one second.
  • Simply aim at the object to be measured and press the button
  • For many uses in household, business and hobby
  • Waterproof IP 65 (FlashStick)