Digital thermo-hygrometer with external sensor
Lieferant: TFA DOSTMANN
5020-5013
DOST5020-5013EA 103 EUR
DOST5020-5013
Digital thermo-hygrometer with external sensor
Hygrometer Thermohygrometer, digital
These instruments are made with MAX-MIN function and external temperature/​humidity sensor (1,5 m cable length).
  • Very fast humidity sensor
  • C/F switchable
Used for table or wall­mounting. With the alarm function (acoustic and optic alarm), it is the ideal solution for laboratories, production plants, green­houses.