MOLYKOTE® 111 Compound Valve Lubricant and Sealant
Lieferant: DOW CORNING
23900034
DOWC23900034EA 40.9 EUR
DOWC23900034
MOLYKOTE® 111 Compound Valve Lubricant and Sealant
Schmiermittel
Lubrication for control and pressure plug valves, water softener and faucet valves. Sealant for vacuum and pressure systems and outdoor equipment (also shipboard) subject to washing and harsh environmental exposure, such as meters, electrical service entrance and underground connections.
  • Water resistance
  • Wide service-temperature range
  • Compatible with many plastics and elastomers
  • Low vapor pressure and volatility
Damping medium for dash pots in electrical and electronic equipment. As an anti-stick and a sealant for transformer gasket and equipment enclosures – it prevents gaskets from sticking to metal and resists weathering and water washout. Also suitable for rubber and plastic O-rings, gaskets and seals.
Standards: NLGI grade 3-4.