Lieferant: DOW CORNING
25100525
DOWC25100525EA 35.1 EUR
DOWC25100525
Klebstoffe
A grease-like silicone material, thickened with heat conductive metal oxides.
  • High thermal conductivity with low bleed
  • No drying, hardening or melting
  • Stable at high temperatures
  • Suitable for the base and mounting studs of transistors, diodes and effective many heat sink devices
Conductivity: 0,67W/m.k
Cure time: 24 h