Silikonelastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Lieferant: DOW CORNING
42417.2
DOWC42417.2EA
0
EUR
DOWC42417.2
Silikonelastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Silikonelastomer
Dow Corning® 3-4241 Dielectric Tough Gel Clear-Green is a two component, heat curing, silicone gel that is used for electrical insulation and protecting electrical components from contaminants, moisture, and stress.
- Fast heat cure
- Two parts are blue and yellow and turn green when mixed (1:1)
- Conditional primerless adhesion at room temperature
- Mechanical strength
- UL 94 V-1 Flammability Rating
- UV indicator for inspection
- Long working time