Gas detection tubes for alcohols
Lieferant: Dräger
8103761
DRAE8103761EA 106 EUR
DRAE8103761
Gas detection tubes for alcohols
Gasrohre
To measure the 'spot concentrations' of alcohol for use with an Accuro pump.
Cross sensitivity Alcohol 100/a: The tube does not differentiate between different alcohols. Higher molecular weight alcohols are indicated with a markedly reduced sensitivity. Aldehydes, ethers, ketones, esters are indicated but with different sensitivity. Aromates, petroleum, halogenated hydrocarbons are not indicated.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.