Pipette controller, Portable Pipet-Aid®
Lieferant: DRUMMOND
4-000-200
DRUM4-000-200EA 1030 EUR
DRUM4-000-200
Pipette controller, Portable Pipet-Aid®
Pipetten Pipettierhelfer
The Pipet-Aid® was designed to transfer liquids with the use of serological, volumetric and/or blow-out pipettes in either glass, plastic or something similar. It accepts 1 to 100 ml pipettes.
  • Filtered nosepiece provides positive protection for the electronic components and prevents cross-contamination due to over-pipetting
  • Pipettes over 2000 times without recharging
  • Flow rate better than 25 ml at 5 s with 100 ml pipette
  • Cordless convenience
  • Tissue Culture Nosepiece provides quick disconnect and easier assembly
  • Holster-type wall bracket included
Lieferumfang: Supplied with charger, Ni-MH rechargeable battery, four replacement filters and wall bracket.