Pipette controller, Pipet-Aid® Hood Mate®
Lieferant: DRUMMOND
4-000-302
DRUM4-000-302EA 1360 EUR
DRUM4-000-302
Pipette controller, Pipet-Aid® Hood Mate®
Pipetten Pipettierhelfer
This controller is specifically designed for convenient, comfortable pipetting under a hood. Ideal for transfering liquids with the use of serological, volumetric and/or blow-out pipettes in either glass, plastic or something similar.
  • Longer coiled hose
  • Lightweight and manufactured from UV resistant materials
  • Extremely lightweight handle provides nine fill and empty speed settings and proportional fingertip control
  • Handle holder incorporates 'auto-off' feature: automatically shuts unit off after 3 m of non-use
  • Dedicated pump housing attaches to hood wall which saves space and prevents spills
  • Coiled vacuum/pressure line entering handle from the top prevents accidental knocking over of bench items