Storage Cabinets for Non-flammable Media
Lieferant: DUPERTHAL
20-200967-050
DUEH20-200967-050EA 867.44 EUR
DUEH20-200967-050
Storage Cabinets for Non-flammable Media
Möbel Lagerschränke
The storage of solid and liquid, non-flammable and non-corrosive chemicals and is specifically marked for the storage of toxic substances.
  • Sheet steel cabinet with hinged doors, painted in RAL 7004 (signal gray)
  • Equipment: Hinged doors with twist handle security lock, closing on 3 sides for safe access
  • Floor collecting tray made of galvanized sheet steel according to StawaR
  • 3x perforated storage floor made of galvanized sheet steel
  • Height adjustable with anti-lift device, Adjustable feet for level adjustment
Standards: Type testing by TÜV SÜD according to furniture standards DIN EN 16121 and DIN EN 16122.