Safety Cabinets for Storage of Flammable Liquids, Type 90, COMPACT line
Lieferant: DUPERTHAL
29-201777-011 29-200977-031
DUEH29-201777-011EA 6228.36 EUR
DUEH29-201777-011 DUEH29-200977-031
Safety Cabinets for Storage of Flammable Liquids, Type 90, COMPACT line
Möbel Lagerschränke
Classical door technology (door handle) for the storage of flammable liquids type testing by MPA/TÜV SÜD according to DIN EN 14470-1, DIN EN 16121 and DIN EN 16122, EK5/AK4 09-10:2009 and AfPS GS 2014: 01 PAK mark of approval: GS/CE mark, high quality mark of approval by TÜV SÜD construction and colour.
  • Outer carcass sheet steel, RAL 7035
  • Doors sheet steel, RAL 1018
  • Inner carcass of decor panels, RAL 7035
  • Doors close automatically in case of fire
  • Doors remain open in every position
  • Cylinder lock for safe access
  • Bottom tray sheet steel, RAL 7035
  • Hot-dip galvanised grating
Exhaust air connection socket on the cabinet roof. Visual inspection of the ventilation opening. Earthing connection on the cabinet roof. Interior fittings conductively connected with carcass. Adjustable foot for level adjustment. Set-back base, RAL 7015.