Safety Cabinets, Type 90, with Wing Door Technology, CLASSIC LINE PURE
Lieferant: DUPERTHAL
29-200667-006 29-201267-006
DUEH29-200667-006EA 6680.74 EUR
DUEH29-200667-006 DUEH29-201267-006
Safety Cabinets, Type 90, with Wing Door Technology, CLASSIC LINE PURE
Möbel Lagerschränke
Classical door technology (door handle), door hung right for the storage of flammable liquids.
  • Outer carcass sheet steel, silver
  • Door sheet steel, silver
  • Inner carcass, stainless steel 1.4301, bottom tray, stainless steel 1.4301
  • Silver similar to RAL 9006
  • Door closes automatically in case of fire, door remains open in every position
  • Cylinder lock for safe access
  • 4× storage shelves, stainless steel 1.4301
  • Exhaust air connection socket on the cabinet roof
Visual inspection of the ventilation opening. Earthing connection on the cabinet roof. interior fittings conductively connected with carcass. Adjustable foot for level adjustment.
Standards: Type testing by MPA/TÜV SÜD according to DIN EN 14470-1, DIN EN 16121 and DIN EN 16122, EK5/AK4 09-10:2009 and AfPS GS 2014: 01 PAK mark of approval: GS/CE mark, high quality mark of approval by TÜV SÜD construction.