Cleanroom sleeve protectors, Tyvek® IsoClean®, Model IC501B WS
Lieferant: DuPont
IC501BWHWS
DUPOIC501BWHWSEA 201 EUR
DUPOIC501BWHWS
Cleanroom sleeve protectors, Tyvek® IsoClean®, Model IC501B WS
Einweg-Kleidung
These sleeves, model IC501BWH WS, are made of Tyvek®, which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Ultra-low linting fabric
  • Bound internal seems
  • Tunnelled elastic at wrist and bicep
  • 450 mm length
  • Aseptically folded
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments and accessories made of sterile Tyvek® IsoClean® are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments.
Standards: Partial body chemical protective clothing, Type PB [6-B], EN 14126, Cat. III
Verpackung: 100 per box, individually packed in pairs. Subgrouped by 25 in an outer bag. Two polyethylene liners. Cardboard box.