Cleanroom hoods, Tyvek® IsoClean®, model IC668B 00, non sterile
Lieferant: DuPont
IC668BWH00
DUPOIC668BWH00EA 324 EUR
DUPOIC668BWH00
Cleanroom hoods, Tyvek® IsoClean®, model IC668B 00, non sterile
Einweg-Kleidung
This hood is not clean-processed and not sterilised made of Tyvek®, a flash spun high density polyethylene. The fabric is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size) and is ultra-low linting.
  • Full face opening with loops and ties
  • Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
  • Partial body chemical protective clothing
  • Barrier to infective agents
Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms. Ideal for use in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments.
Standards: Certified according to Regulation (EU) 2016/425; Partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
Verpackung: Packed in cardboard box, 100 per box, bulk packed. 2 polyethylene liners.