Cleanroom floor mats, floating systems, CleanZone
Lieferant: DYCEM
CZ01FS1206 CZ01FS1210 CZ01FS1220 CZ01FS2006 CZ01FS2010 CZ01FS2020 CZ01FS2030 CZ01FS2040 CZ01FS2350 CZ01FS2010MN CZ01FS1225M CZ01FS1206MN
DYCECZ01FS1206EA 2320 EUR
DYCECZ01FS1206 DYCECZ01FS1210 DYCECZ01FS1220 DYCECZ01FS2006 DYCECZ01FS2010 DYCECZ01FS2020 DYCECZ01FS2030 DYCECZ01FS2040 DYCECZ01FS2350 DYCECZ01FS2010MN DYCECZ01FS1225M DYCECZ01FS1206MN
Cleanroom floor mats, floating systems, CleanZone
Fußmatten Permanente Bodenmatten
These floating floor mats are high performance, antimicrobial loose-laid mats offering many benefits without the need for permanent installation. A floating floor can be placed over smooth or rough subfloors and can be self-installed with ease.
  • Reduced floor (up to 99,9%) and air particulate (up to 75%) counts in controlled or critical areas
  • Long lasting, cost effective solution compared with peel-off mats and other alternatives
  • Reduced microbial counts due to Biomaster, a high performance anti-microbial additive
  • Easy to clean, maintain, and incorporate
More Product Information