Alcohol metabolite Standards, ethyl sulphate sodium salt, CRM, Cerilliant®, Supelco®
Lieferant: Merck
E-064-1ML
E-066-1ML
E-116-1ML
E-064-1MLEA
181
EUR
E-064-1ML
E-066-1ML
E-116-1ML
Alcohol metabolite Standards, ethyl sulphate sodium salt, CRM, Cerilliant®, Supelco®
Standards
Organische Standards
Ethyl sulfate is a primary urinary metabolite of consumed alcohol and is indicative of recent alcohol consumption.
- Snap-N-Spike®/Snap-N-Shoot®
Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine alcohol testing.
Bitte beachten Storage temperature: −20 °C (freezer).