Ethidium homodimer I 2 mM in DMSO ≥95% (durch TLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: EthD-1, EtDi

M-Clarity™ Qualitätsstufe = MQ100

E1903-.5ML
E1903-.5MLEA 372 EUR
E1903-.5ML
Ethidium homodimer I 2 mM in DMSO ≥95% (durch TLC), Sigma-Aldrich®
Ethidium homodimer I

Ethidium homodimer is a red fluorescent dye. It cannot cross an intact plasma membrane, but can easily pass through a damaged plasma membrane. Ethidium homodimer is used to check membrane integrity. One molecule of EthD-1 will non-selectively bind four base pairs in dsDNA.


  • Ethidium homodimer I solution has been used to check membrane integrity in sperms
  • Stains ssDNA, dsDNA, RNA, oligonucleotides, and triplex DNA structures
  • Used to detect DNA in solution

Formel: C₄₆H₅₀Cl₄N₈
Molecular Weight: 856.75 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00210948
CAS-Nummer: 61926-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abc3c2e3b3a77bb44fa5dfc07b280559bc05a80&vid=63cc11624ca6e92b2aefa9e063af93ac42c44f2d&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Red
Appearance (Form) Liquid
E
Lambda Max at 490-496, in H2O
Run on Ethidium Homodimer before dissolution in DMSO
≥9000
Wavelength
Lambda Max
490 - 496 nm
Purity (TLC)
Run on Ethidium Homodimer before dissolution in DMSO
≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR