Evans-Blau ≥75% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

E2129-100G E2129-10G E2129-50G
E2129-100GEA 418 EUR
E2129-100G E2129-10G E2129-50G
Evans-Blau ≥75% (Farbstoffgehalt), Sigma-Aldrich®
Evans blue

Evans blue has been used to check cell viability. It has been used to study BBB permeability.

Formel: C₃₄H₂₈N₆O₁₄S₄
Lagertemperatur: Room Temperature
MDL: MFCD00004021
CAS-Nummer: 314-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c925c923a3ac1be786570777bd28fd52598f8ebb&vid=ac73f772d8c8a7c292398f4744714cffe6024ef0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder or Crystals or Granules
Carbon 30.0 - 42.5 %
Nitrogen 6.1 - 8.7 %
Dye Content ≥ 75 %
Recommended Retest Period 4 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR