Eosin G, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 2,4,5,7-Tetrabromfluorescein Dinatriumsalz, Tetrabromfluorescein Dinatriumsalz, Solvent Red 43, Dinatrium-2-(2,4,5,7-tetrabrom-6-oxido-3-oxoxanthen-9-yl)benzoat, 2′,4′,5′,7′-Tetrabrom-3′,6′-dihydroxyspiro [2-benzofuran-3,9′-xanthen]-1-on Dinatriumsalz, Eosin 225, Eosin gelbstichig, Eosin G Dinatriumsalz, Eosin gelblich, Eosin Y Dinatriumsalz, Eosin Y

M-Clarity™ Qualitätsstufe = MQ200

E6003-100G E6003-250G E6003-25G
E6003-100GEA 74 EUR
E6003-100G E6003-250G E6003-25G
Eosin G, Sigma-Aldrich®
Eosin G

Dye content - ≥85%.


Eosin Y disodium salt has been used for determining sperm viability. The swollen and stained spermatozoa were considered dead. In corneal epithelium, it has been used in the procedure of immunohistological staining. Eosin Y is a xanthene dye and is used for the differential staining of connective tissue and cytoplasm. In histopathology, it is applied as a counterstain after hematoxylin and before methylene blue. It is also used as a background stain, thereby giving contrast to the nuclear stains.

Formel: C₂₀H₆Br₄Na₂O₅
Schmelzpunkt: 255,..270 °C
Dichte: 1,018 g/cm³
Flammpunkt: 191,6 °C
Lagertemperatur: Room Temperature
MDL: MFCD00005040
CAS-Nummer: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7962fedb3666fca5d6418ac691ec4eb29d7866bd&vid=918729b2b9d73f40ae38dcb4d39d404fcb2113a5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Solubility (Colour) 1 mg/ml H₂O Orange to Very Dark Orange and Red to Very Dark Red and Orange-Red and Red-Orange
Solubility (Turbidity) Clear
Dye Content ≥85%
pH 6.55 (29 °C, 84 °F)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR