PBS-TWEEN pH 7.5, ultrarein
Lieferant: VWR Chemicals
E715-1LEA
229
EUR
E715-1L
PBS-TWEEN pH 7.5, ultrarein
Puffer
Elektrophorese-Puffer
Waschpuffer
Phosphate Buffered Saline (PBS) with 0,05% Tween® 20, pH 7,5 is a ready-to-use 1× concentration.