Neutralrot, zertifizierbare Qualität

Lieferant: VWR Chemicals

Synonyme: Basisrot 5, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid

E895-100GEA 990 EUR
E895-100G
Neutralrot, zertifizierbare Qualität
Neutralrot

A cationic azine dye slightly soluble in water and ethanol. Used as a weak nuclear stain and as a supravital stain of fresh peripheal blood.

Formel: C₁₅H₁₇ClN₄
Molecular Weight: 288.78 g/mol
Schmelzpunkt: 290
Lagertemperatur: Raumtemperatur
MDL: MFCD00012651
CAS-Nummer: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR