The SL3101 set by ebro, featuring five EBI 12-T471 and one EBI 12-TP453 loggers in silicone protection boxes, is ideal for validating large sterilisers and autoclaves in demanding environments.
- Comprehensive set for large sterilizers and autoclaves
- Includes 5× EBI 12-T471 and 1× EBI 12-TP453 loggers
- Silicone protection boxes (AL 107 and AL 101) for added heat resistans
- Traceable factory calibration certificate
The SL3101 set by ebro is designed for the validation and monitoring of large sterilizers and autoclaves, essential for maintaining high sterilization standards in healthcare and industrial settings. This set includes five EBI 12-T471 temperature loggers and one EBI 12-TP453 combined temperature and pressure logger, each equipped with silicone protection boxes (AL 107 for EBI 12-T471 and AL 101 for EBI 12-TP453) for added safety during high-demand cycles. With precise temperature and pressure measurements, the SL3101 set ensures comprehensive monitoring and supports consistent compliance with sterilization standards.
Dimensions (loggers): Ø 48×24 mm H.
Lieferumfang: Includes 5× EBI 12-T470 Temperature data logger, 1× EBI 12-TP453 pressure and tempreture logger, 1× EBI IF 200 interface, 1× Winlog.validation software, 1× carrying case 'SYSTAINER', manual, traceable factory calibration certificate.