Infrared thermometer, TFI 54
Lieferant: EBRO
1340-1754
EBRO1340-1754EA 110 EUR
EBRO1340-1754
Infrared thermometer, TFI 54
Thermometer Digitalthermometer
Infrared thermometer with splash-proof housing for non-contact surface measurements.
  • Rubberized housing
  • Single laser pointer
  • Optics D:S = 12:1
  • Two fixed emission levels for goods and packaging
  • Replaceable battery
Suitable for logistics, kitchen, serving food and checking goods in frozen stores.