pH tester, PHX 800
Lieferant: EBRO
1340-5800
EBRO1340-5800EA 91 EUR
EBRO1340-5800
pH tester, PHX 800
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
Robust pH tester for use in industrial process water, used water, rinsing water in washer-disinfectors, laboratories, food production and beverage production.
  • Robust and waterproof ABS housing
  • With automatic pH calibration
  • Hold-function, acoustic signal
  • Auto-lock-function
  • Automatic deactivation after 15 min., battery level indicator
  • Exchangeable battery