Robust Temperature Bottle Data Loggers, EBI 12-T26x
Lieferant: EBRO
1340-6618 1340-6619 1340-6620 1340-6621 1340-6622
EBRO1340-6618EA 2550 EUR
EBRO1340-6618 EBRO1340-6619 EBRO1340-6620 EBRO1340-6621 EBRO1340-6622
Robust Temperature Bottle Data Loggers, EBI 12-T26x
Datenlogger
The data logger is specifically designed for monitoring pasteurization processes, particularly in beverage cans or bottles. The sensor needle is inserted externally into the beverage packaging and securely fastened.
  • Wide measuring range, –55 to +140 °C
  • High-quality PT1000 sensor (class A)
  • Rigid, external temperature sensor: Ø 6 mm, various lengths
  • Robust IP 68 protection class: waterproof and vapour-tight
  • M 10 thread on external sensor
The EBI 12-T26x data logger is designed for pasteurization control in beverage cans or bottles. Its Pt 1000 sensor, featuring a blunt, axial probe (M10 thread, Ø 6 mm, various lengths), is inserted and secured into the packaging. Operating within a temperature range of –55 to +140 °C with an accuracy of ±0.3 °C between –40 and +140 °C, this robust logger is ideal for routine quality assurance checks in production. The device comes with factory calibration for dependable performance.
EBI 12-T26x calibration points: –20, +0, +60 and +134 °C
Ø×H: 48×24 mm
Lieferumfang: Includes 1× EBI 12-T26x, 1× factory calibration certificate, manual.