Temperature and Pressure Data Loggers for Process Validation with Flexible Probes, EBI 12-TP451 or -TP453, -TP460
Lieferant: EBRO
1340-6643 1340-6647 1340-6648
EBRO1340-6643EA 6810 EUR
EBRO1340-6643 EBRO1340-6647 EBRO1340-6648
Temperature and Pressure Data Loggers for Process Validation with Flexible Probes, EBI 12-TP451 or -TP453, -TP460
Datenlogger
These data loggers are high-precision temperature and pressure loggers designed for process validation in steam sterilisers, batch control, and pharmaceutical applications. With a wide temperature and pressure range and high accuracy, they provide reliable data logging for critical monitoring processes in sterilisation, healthcare, production, and pharmaceutical environments, ensuring accurate validation and quality control.
  • Suitable for steam sterilisers, process validation
  • Temperature range: 0 to 150 °C
  • High accuracy: up to 0,05 °C
  • Bendable wire probe, Ø
More Product Information